Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiocyanate, 0.100 N (N/10), Ricca Chemical
SDP

Catalog No. 78605 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7860-5 20 L Cubitainer
786016 500 mL Natural Poly Bottle
7860-32 1 L Natural Poly Bottle
7860-25 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 7860-5 Supplier Ricca Chemical Company Supplier No. 78605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 540-72-7 , 7732-18-5
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Color Colorless
CAS Min % 97.0, 0.79
CAS Max % 100.0, 0.82
Grade Laboratory
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 0.100Normal
Chemical Name or Material Sodium Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.