Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium thiocyanate

Catalog No. 5000031039 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-000-31039 250 g
46-787-150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-000-31039 Supplier Sigma Aldrich Supplier No. 467871250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium thiocyanate

Specifications

Melting Point 287°C (lit.)
CAS 540-72-7
Quantity 250 g
Molecular Formula CNNaS
Linear Formula NaSCN
MDL Number MFCD00011123
Synonym Sodium isothiocyanate; Sodium rhodanate; Sodium rhodanide; Sodium sulfocyanate
Molecular Weight (g/mol) 81.07
Percent Purity ≥99.99% (trace metals basis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.