Learn More
Medchemexpress LLC Sodium thiocyanate, GR | 540-72-7 | 99.1% | 81.07 | 50 G
SDP
Catalog No. 5000400533
Encompass_Preferred
Sodium thiocyanate reduces plasma levels of the pro-inflammatory cytokine IL-6 and increases the anti-inflammatory cytokine IL-10 levels. It also significantly reduces ROS formation.
  • Reduces pro-inflammatory cytokine IL-6
  • Increases anti-inflammatory cytokine IL-10
  • Significantly reduces ROS formation
Catalog No. 50-004-00533 Supplier Medchemexpress LLC Supplier No. HY2311950G
Sodium thiocyanate reduces plasma levels of the pro-inflammatory cytokine IL-6 and increases the anti-inflammatory cytokine IL-10 levels. It also significantly reduces ROS formation.
  • Reduces pro-inflammatory cytokine IL-6
  • Increases anti-inflammatory cytokine IL-10
  • Significantly reduces ROS formation