Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thioglycolate, Powder, 98%, Spectrum™ Chemical
SDP

Catalog No. 18613914 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
10 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-914 2.5 kg Amber Glass Bottle
18-613-909 25 g Amber Glass Bottle
18-613-910 100 g Amber Glass Bottle
18-613-912 500 g Amber Glass Bottle
18-613-911 10 kg Amber Glass Bottle
18-613-913 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-613-914 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO2092.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 367-51-1

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 2.5 kg
Molecular Weight (g/mol) 114.09
Percent Purity 98%
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Thioglycolate, Powder

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.