Learn More
Aqua Solutions SODIUM THIOSOLFATE .01N 1PT
SDP
Catalog No. NC9794588
Encompass_Preferred
Sodium Thiosulfate 0.01 Normal Aqueous Solution, N.i.s.t. Traceable (500ml)
Catalog No. NC9794588 Supplier Aqua Solutions Supplier No. 8790500ML
Add to Cart
Add to Cart
Sodium Thiosulfate 0.01 Normal Aqueous Solution, N.i.s.t. Traceable (500ml)