Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.00387N Standardized, Reagents

Catalog No. 01333549
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-549 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-549 Supplier Reagents Holdings Llc Supplier No. CS186100500A
 Restricted Item
Add to Cart
Add to Cart

Sodium Thiosulfate, 0.00387N Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Thiosulfate, 0.00387N Standardized
Boiling Point 100°C
CAS 7732-18-5, 10102-17-7, 497-19-8
Color Colorless
Physical Form Liquid
Quantity 500 mL
Molecular Formula Na2S2O3/H2O
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0007
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.