Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Sodium Thiosulfate, 0.011N
SDPSCI_ED

Catalog No. S72383 Shop All LaMotte Company Products
Change view
Click to view available options
Volume (Metric):
60 mL
3800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S72383 60 mL
S72316 3800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S72383 Supplier LaMotte™ Supplier No. 3819H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Thiosulfate, 0.011N is a replacement reagent.

Available in multiple sizes:

  • Gallon size for multiple tests.
  • Kit size bottle.

Specifications

Color Pale yellow
Composition 10102-17-7, 497-19-8
Concentration 0.011 N
Description Sodium thiosulfate, Sodium carbonate
Volume (Metric) 60 mL
Format Liquid
Type Sodium Thiosulfate
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.