Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.0200 N (N/50), Ricca Chemical
SDP

Catalog No. R78950004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
Cubitainer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R78950004C 4 L Amber Glass Bottle
R78950004F 4 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R78950004C Supplier Ricca Chemical Company Supplier No. 78951
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10102-17-7 , 497-19-8 , 7732-18-5
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Color Colorless
Assay Percent Range 0.02000 ± 0.00001 N at 20°C
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless to slight solvent odor
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Sodium Thiosulfate, 0.0200 Normal (N/50)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.