Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.0375 N, Ricca Chemical
SDP

Catalog No. 79255 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7925-5 20 L Cubitainer
7925-32 1 L Natural Poly Bottle
7925-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7925-5 Supplier Ricca Chemical Company Supplier No. 79255
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Color Colorless
CAS Min % 0.73, 97.0, 0.01
CAS Max % 0.76, 100.0, 0.01
Grade Laboratory
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 0.0375Normal
Chemical Name or Material Sodium Thiosulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.