Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.050 Normal (N/20), Reagents

Catalog No. 01333971
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-971 4 L
01-333-973 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-971 Supplier Reagents Holdings Llc Supplier No. CS0100004C
Only null left
Add to Cart
Add to Cart

Sodium Thiosulfate, 0.050 Normal (N/20), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 10102-17-7 , 497-19-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0053
Chemical Name or Material Sodium Thiosulfate, 0.050 Normal (N/20)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.