Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.1 N (N/10) Standard Volumetric Solution, Macron Fine Chemicals™

Catalog No. MK46821 Shop All Avantor Products
Change view
Click to view available options
1L; Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-4682-1 1L; Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-4682-1 Supplier Avantor Supplier No. 468260
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7772-98-7
Physical Form Liquid
Quantity 1 L
Molecular Formula Na2O3S2
MDL Number MFCD00003499
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium sulfanidesulfonate
Molecular Weight (g/mol) 158.10
PubChem CID 24477
Appearance Clear, colorless solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.