Learn More
Hach Company Sodium Thiosulfate Digital Titrator Cartridge, 0.113 N
Supplier:  Hach Company 2267301
Encompass
For chlorine determination by iodometric Digital Titrator titration. 0.113 N, Digits x 0.1 = mg/L CI2 in a 5 mL sample, Digits x 0.000141 = meq. Approximately 40-100 tests
Catalog No. 50-199-5682