Learn More
Hach Company Sodium Thiosulfate Digital Titrator Cartridge, 0.200 N
Supplier:  Hach Company 2267501
Encompass
For determination of Dissolved Oxygen by Winkler Method Digital Titrator titration. 0.200 N Na2S2O3. Digits x 0.01 = mg/L DO in 200 mL sample, Digits x 0.00025 = meq.
Catalog No. 50-199-5684