Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.500 N (N/2), Ricca Chemical
SDP

Catalog No. 79665 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
79665 20 L Cubitainer
7966-16 500 mL Natural Poly Bottle
7966-32 1 L Natural Poly Bottle
7966-1 4 L Natural Poly Bottle
796625 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 79665 Supplier Ricca Chemical Company Supplier No. 79665
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 51-98-9, 10102-17-8, 497-19-9
Molecular Formula C22H28O3
Synonym norethisterone acetate, 19-norethindrone acetate, norethindrone acetate, enta, orlutate, norlutin acetate, norlutate, 19-norethisterone acetate, primolut-nor, aygestin
InChI Key IMONTRJLAWHYGT-ZCPXKWAGSA-N
SMILES CC(=O)OC1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34)C)C#C
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 340.463
PubChem CID 5832
ChEBI CHEBI:7628
Grade Laboratory
Packaging Cubitainer
pH ∼10
Quantity 20 L
Chemical Name or Material Sodium thiosulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.