Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 1.00 N, Ricca Chemical
SDP

Catalog No. R7970000250 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R7970000250 250 mL Natural Poly Bottle
7970-16 500 mL Natural Poly Bottle
7970-32 1 L Natural Poly Bottle
R79700004F 4 L Cubitainer
7970-1 4 L Natural Poly Bottle
797025 10 L Cubitainer
7970-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. R7970000250 Supplier Ricca Chemical Company Supplier No. 79708
Add to Cart
Add to Cart

Oxidation-Reduction Titrant

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Color Colorless
Boiling Point ∼100°C
CAS Min % 22.12, 77.7, 0.18
Melting Point ∼0°C
CAS Max % 22.12, 77.7, 0.18
Density 1.13
Solubility Information Miscible in water
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼10
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Sodium Thiosulfate, 1.00 Normal
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.