Learn More
Hach Company Sodium Thiosulfate Digital Titrator Cartridge, 2.00 N
Supplier:  Hach Company 1440101
Encompass
For determination of Dissolved Oxygen by the Winkler method and Digital Titrator titration. 2.0 N. Digits x 0.1 = mg/L DO in 200 mL sample. Digits x 0.00025 = meq.
Catalog No. 50-199-5025