Learn More
Hach Company Sodium Thiosulfate Solution, 2.12N 100 mL
Supplier:  Hach Company 2686832
Encompass
For determination of high range hypochlorite (bleach) concentrations by drop-count titration
Catalog No. 50-199-6542