Learn More
Electron Microscopy Sciences Sodium Thiosulfate 5% Aqueous 500 ML
SDP
Catalog No. 5031770
Encompass_Preferred
Used in Lillie Modification of Masson's Trichrome for staining of cells, cytoplasm muscle and collagen of mammalian tissue. See references: Clark, G.: Staining Procedures. Williams & Wilkins Co., Baltimore, 3rd ed., c. 1973, p.59 and Lillie, R.D., Arch. Path., 29:705, 1940
Catalog No. 50-317-70 Supplier Electron Microscopy Sciences Supplier No. 2636408
Used in Lillie Modification of Masson's Trichrome for staining of cells, cytoplasm muscle and collagen of mammalian tissue. See references: Clark, G.: Staining Procedures. Williams & Wilkins Co., Baltimore, 3rd ed., c. 1973, p.59 and Lillie, R.D., Arch. Path., 29:705, 1940