Learn More
Electron Microscopy Sciences Sodium Thiosulfate 5% Aqueous 500 ML
SDP
Catalog No. 5031833
Encompass_Preferred
Used in Methenamine Silver Methodfor Argentaffin Cells for staining of argentaffin granules of enterochromaffin cells. See Clark, G.: Staining Procedures, Williams and Wilkins Co., Baltimore, 3rd Ed., c. 1973, p. 161, Gomori, G., Arch. Path., 45:48, 1948 or Burtner, H.J. and Lillie, R.D., Stain Techn., 24:225-7, 1949.
Catalog No. 50-318-33 Supplier Electron Microscopy Sciences Supplier No. 2648102
Used in Methenamine Silver Methodfor Argentaffin Cells for staining of argentaffin granules of enterochromaffin cells. See Clark, G.: Staining Procedures, Williams and Wilkins Co., Baltimore, 3rd Ed., c. 1973, p. 161, Gomori, G., Arch. Path., 45:48, 1948 or Burtner, H.J. and Lillie, R.D., Stain Techn., 24:225-7, 1949.