Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 8% (w/v), Ricca Chemical
SDP

Catalog No. R78672001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R78672001A 1 L Natural Poly Bottle
R78672004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R78672001A Supplier Ricca Chemical Company Supplier No. R78672001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Color Colorless
CAS Min % 91.0, 7.26
CAS Max % 94.0, 7.56
Percent Purity 8%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 8% (w/v)
Chemical Name or Material Sodium Thiosulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.