Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 0.93, 99.05, 0.02 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Linear Formula | Na2S2O3·5H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 248.18 |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 0.0375N ±0.0003N (0.0375M) |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.