Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Pentahydrate, ACS Reagent, Reagents

Catalog No. p-200020916
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-661 10 kg
01-337-659 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-661 Supplier Reagents Holdings Llc Supplier No. C132800010KG
Add to Cart
Add to Cart

Sodium Thiosulfate Pentahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Boiling Point 100°C
Melting Point 48°C
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 1.69
Chemical Name or Material Sodium Thiosulfate Pentahydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.