Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, Pentahydrate, Crystal, USP, EP, BP, JP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606899 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-899 50 kg Fiber Drum
18-606-896 500 g Poly Bottle
18-606-897 2.5 kg Poly Bottle
18-606-898 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-899 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO21050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO210, 10102-17-7, Na2S2O3·5H2O

Spectrum™ Chemical Sodium Thiosulfate, Pentahydrate, Crystal, USP, EP, BP, JP is used to treat cyanide poisoning and pityriasis versicolor. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

pH 6.0 to 8.0
CAS Max % 1
Quantity 50 kg
Percent Purity 99 to 100.5%
Grade Multi-Compendial
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.