Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Pentahydrate, For Analysis, EMSURE™, ACS, MilliporeSigma™

Catalog No. M1065169025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065169025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065169025 Supplier MilliporeSigma Supplier No. 1065169025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Melting Point 48°C
Color Colorless
pH 6.0 to 7.5 (100g/L, H2O, 20°C)
Physical Form Solid
Quantity 25 kg
Solubility Information 701g/L
Formula Weight 248.21
Percent Purity 99.5 to 101.0%
Grade ACS
Packaging Fiber Carton
Density 1.74 g/3 (20 °C)
Show More Show Less
EINECSNumber 231-867-5
RTECSNumber WE6660000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.