Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 100°C |
| Melting Point | 0°C |
| Concentration | 0.1995 to 0.2005 |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Traceability to NIST | Traceable to NIST. |
| Vapor Pressure | 14mmHg |
| Quantity | 4 L |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. NFPA Health:1 Flammability:0 Instability:0 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?