Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Solution, (0.1 N) Titripur™ Reag. Ph Eur,Reag. USP; MilliporeSigma™ Sodium Thiosulfate Solution, (0.1 N) Titripur™ Reag. Ph Eur,Reag. USP; MilliporeSigma™

Catalog No. M1091474000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Titripac PE/Alu
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1091474000 4 L Titripac PE/Alu
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1091474000 Supplier MilliporeSigma Supplier No. 1.09147.4000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Physical Form Liquid
Quantity 4 L
Grade Reagent Ph. Eur./Reagent USP
Packaging Titripac PE/Alu
Density 1.01g/cm3 (20°C)
Chemical Name or Material Sodium Thiosulfate Solution
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.