Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Solution, 100 mL, G-Biosciences
SCI_ED

Catalog No. S001110
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001110 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001110 Supplier G-Biosciences Supplier No. 7862243
Add to Cart
Add to Cart

Sodium Thiosulfate Solution is used in the Silver Stain Kit.

  • It is also used in water treatment and to neutralize bleach.

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Sodium Thiosulfate
Quantity 100 mL
Solution Type Buffer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.