Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, Solution, Titripur™, Reagent grade;,MilliporeSigma™,

Catalog No. M1091471000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1091471000 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1091471000 Supplier MilliporeSigma Supplier No. 1.09147.1000
Add to Cart
Add to Cart

Recommended Storage

+15°C to +25°C.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
pH 9 to 10 (H2O, 20°C)
Physical Form Liquid
Quantity 1 L
Solubility Information Soluble
Packaging Plastic Bottle
Density 1.01 g/3 (20 °C)
Recommended Storage +15°C to +25°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.