Learn More
Hach Company Sodium Thiosulfate Digital Titrator Cartridge, Stabilized, 0.02256 N
Catalog No. 501995935 Shop All Hach Company Products
Encompass
For determination of Chlorine by Digital Titrator titration. 0.02256 N Digits x 0.01 = mg/L CI2 in 100 mL, Digits x 0.0000282 = meq. Stabilized sodium thiosulfate formulation.
Catalog No. 50-199-5935 Supplier Hach Company Supplier No. 2409101
For determination of Chlorine by Digital Titrator titration. 0.02256 N Digits x 0.01 = mg/L CI2 in 100 mL, Digits x 0.0000282 = meq. Stabilized sodium thiosulfate formulation.