Learn More
Hach Company Sodium Thiosulfate Standard Solution, Stabilized, 0.0246 N, 100 mL MDB
Catalog No. 501995936 Shop All Hach Company Products
Encompass
Stabilized sodium thiosulfate titrant for chlorine determination by iodometric titration. 100 mL Marked Dropping Bottle
Catalog No. 50-199-5936 Supplier Hach Company Supplier No. 2409232
Stabilized sodium thiosulfate titrant for chlorine determination by iodometric titration. 100 mL Marked Dropping Bottle