Learn More
Hach Company Sodium Thiosulfate Digital Titrator Cartridge, Stabilized, 0.0250 N
Catalog No. 501995938 Shop All Hach Company Products
Encompass
For dissolved oxygen determination by Winkler titration. Digits x 0.1 = mg/L DO in 25 mL. Stabilized sodium thiosulfate formulation
Catalog No. 50-199-5938 Supplier Hach Company Supplier No. 2409301
For dissolved oxygen determination by Winkler titration. Digits x 0.1 = mg/L DO in 25 mL. Stabilized sodium thiosulfate formulation