Learn More
Hach Company Sodium Thiosulfate (Stabilized) Digital Titrator Cartridge, 2.26 N
Catalog No. 501996543 Shop All Hach Company Products
Encompass
For determination of Hypochlorite (Bleach) by Digital Titrator titration. Hach Method 10100. Range: 5 - 15% Cl2 (50 - 150 g/L Cl2). Stabilized sodium thiosulfate formulation. 2.26 N Digits x 0.5 = g/L CI2 in 0.2 mL sample
Catalog No. 50-199-6543 Supplier Hach Company Supplier No. 2686901
For determination of Hypochlorite (Bleach) by Digital Titrator titration. Hach Method 10100. Range: 5 - 15% Cl2 (50 - 150 g/L Cl2). Stabilized sodium thiosulfate formulation. 2.26 N Digits x 0.5 = g/L CI2 in 0.2 mL sample