Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium triacetoxyborohydride, 97%

Catalog No. ac291820250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291820250 25 g Glass bottle
AC291821000 100 g Glass bottle
AC291825000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC291820250 Supplier Thermo Scientific Chemicals Supplier No. 291820250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 56553-60-7 | C6H10BNaO6 | 211.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56553-60-7
Molecular Formula C6H10BNaO6
Molecular Weight (g/mol) 211.94
MDL Number MFCD00012211
InChI Key HHYFEYBWNZJVFQ-UHFFFAOYSA-N
Synonym c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
IUPAC Name sodium bis(acetyloxy)boranuidyl acetate
SMILES [Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Melting Point 114°C to 118°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Linear Formula NaB(O2CCH3)3H
Quantity 25 g
Fieser 12,453; 13,283; 06,553; 16,309
Merck Index 15, 8822
Solubility Information Solubility in water: reacts. Other solubilities: soluble in thf,acetonitrile,1,2-dichloroethane
Formula Weight 211.94
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Sodium triacetoxyborohydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.