Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Trichloroacetate

Catalog No. 190780100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-078-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 19-078-0100G Supplier Sigma Aldrich Supplier No. 190780100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium trichloroacetate with trichloroacetic acid (TCA) undergoes rapid decarboxylation in dimethylformamide (DMF) in the presence of aldehydes to form nucleophilic trichloromethyl anion, which then adds to the aldehydes to form trichloromethyl carbinols. It can also undergo thermal decarboxylation in aprotonic solvents in the presence of olefins to generate dichlorocarbene as an intermediate.

Specifications

CAS 7787-20-4
Melting Point 5°C to 6°C
Density 0.948 g/mL at 25°C
Boiling Point 192°C to 194°C
Molecular Formula C10H16O
Linear Formula C10H16O
Quantity 100 g
MDL Number MFCD00151104
Synonym NaTCA; TCA sodium salt; Trichloroacetic acid sodium salt
Molecular Weight (g/mol) 152.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.