Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Triethylborohydride Solution

Catalog No. 227307100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-730-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 22-730-7100ML Supplier Sigma Aldrich Supplier No. 227307100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of alcohols by hydrogenation of ketones catalyzed by an iron pincer complexPreparation of alkali metal boron monohydride complexesCoordinative substitution reactionsLigand exchange reactionsHydridic reagent

Specifications

CAS 4551-15-9
Density 0.963 g/mL at 25°C
Boiling Point 72°C (8 mmHg, lit.)
Molecular Formula C9H14SSi
Linear Formula C6H5SSi(CH3)3
Quantity 100 mL
MDL Number MFCD00008272
Synonym Sodium triethylborate; Sodium triethylhydridoborate; Sodium triethylhydroborate
Molecular Weight (g/mol) 182.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.