Learn More
Medchemexpress LLC Sodium trifluoroacetate-13C2 | 1794767-05-7 | MFCD22378824 | 99.9% | 137.99 g/mol | 13C2F3NaO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW017049S1MG
Encompass_Preferred
Sodium trifluoroacetate-13C2 is the 13C2 isotopically labeled sodium salt of trifluoroacetic acid, provided as an analytical reference standard for use as an internal standard or tracer in quantitative NMR, GC-MS, and LC-MS analyses. It is intended for analytical and method development applications where precise isotopic labeling improves accuracy and reproducibility.
  • Isotopically labeled with two 13C atoms for traceable mass and NMR shifts.
  • High purity minimizes interference in analytical assays.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in small quantities appropriate for analytical use.
  • Provides consistent performance for quantitative method development.
Catalog No. 50-003-01270