Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Trifluoroacetate, MP Biomedicals

Catalog No. ICN205622 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205622 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205622 Supplier MP Biomedicals, Inc Supplier No. 0520562210

Chemical Identifiers

CAS 2923-18-4
Molecular Formula C2F3NaO2
Molecular Weight (g/mol) 136.01
MDL Number MFCD00013217
InChI Key UYCAUPASBSROMS-UHFFFAOYSA-M
Synonym sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
PubChem CID 517019
IUPAC Name sodium trifluoroacetate
SMILES [Na+].[O-]C(=O)C(F)(F)F

Specifications

Quantity 10 g
Formula Weight 136
Chemical Name or Material Sodium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.