Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Trifluoromethanesulfinate

Catalog No. 7432325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
234 mg
2 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-323-25G 5 g
74-323-2234MG 234 mg
74-323-22G 2 g
74-323-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 74-323-25G Supplier Sigma Aldrich Supplier No. 7432325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium Trifluoromethanesulfinate

Specifications

CAS 2926-29-6
Melting Point <325°C
Molecular Formula CF3NaO2S
Linear Formula CF3NaO2S
MDL Number MFCD03092989
Quantity 5 g
Synonym Langlois reagent; Sodium trifluoromethanesulfinate; Sodium trifluoromethylsulfinate; Trifluoromethanesulfinic acid sodium salt
Molecular Weight (g/mol) 156.06 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.