Learn More
Accela Chembio Inc Sodium Trimethylacetate Hydrate | 25g | 143174-36-1 | MFCD00150782 | 99% | Shelf Life: 1620 Days | Light Sensitive
Catalog No. 50354974
Encompass_Preferred
Sodium Trimethylacetate Hydrate | 25g | 143174-36-1 | MFCD00150782 | 99% | Shelf Life: 1620 Days | Light Sensitive
Catalog No. 50-354-974 Supplier Accela Chembio Inc Supplier No. SY01163525G
Sodium Trimethylacetate Hydrate | 25g | 143174-36-1 | MFCD00150782 | 99% | Shelf Life: 1620 Days | Light Sensitive