Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Triphosphate (Sodium Tripolyphosphate), Reagents

Catalog No. 01336948
Change view
Click to view available options
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-948 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-948 Supplier Reagents Holdings Llc Supplier No. C2328300100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Triphosphate (Sodium Tripolyphosphate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7758-29-4
Molecular Formula Na5O10P3
Molecular Weight (g/mol) 367.86
InChI Key HWGNBUXHKFFFIH-UHFFFAOYSA-I
Synonym Pentasodium Triphosphate
IUPAC Name pentasodium bis(phosphonatooxy)phosphinate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 622°C
Color White
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.5
Chemical Name or Material Sodium Triphosphate (Sodium Tripolyphosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.