Learn More
Electron Microscopy Sciences Sodium Tungstate 25 GR

Supplier: Electron Microscopy Sciences 21400
CAS #10213-10-2 Na₂WO4₂H₂O F.W. 329.86
Assay: 99.0-101.0%
Stockert,J.C. (1977). Sodium Tungstate as a stain in electron microscopy. Biol. Cellul., 29:211. Takeuchi, I.K. (1981). Differential staining of nucleoli and chromatin by sodium tungstate. J. Electron Microsc., 30:15.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.