Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium tungstate dihydrate100G
Catalog No. 502898540
Encompass
Sodium tungstate dihydrate is a hydrated sodium salt of tungstic acid. It participates as a catalyst in the preparation of various epoxides. Various crystallographic properties of sodium tungstate dihydrate have been analyzed.
Catalog No. 50-289-8540 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 223336100G
Sodium tungstate dihydrate is a hydrated sodium salt of tungstic acid. It participates as a catalyst in the preparation of various epoxides. Various crystallographic properties of sodium tungstate dihydrate have been analyzed.