Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium tungstate dihydrate500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 223336500G
Encompass
Sodium tungstate dihydrate is a hydrated sodium salt of tungstic acid. It participates as a catalyst in the preparation of various epoxides. Various crystallographic properties of sodium tungstate dihydrate have been analyzed.
Catalog No. 50-289-8660