Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium valproate | 1069-66-5 | MFCD00078604 | 99.7% | 166.19 g/mol | C8H15NaO2 | 25 G
SDP

Catalog No. 5000244237
Encompass_Preferred

Valproic acid sodium (sodium valproate) is the sodium salt of valproic acid for research use. It acts as an HDAC inhibitor and anticonvulsant, and is commonly used in studies of epilepsy, bipolar disorder, neuroprotection, and epigenetic regulation. Supplied as a solid powder suitable for dissolution for chemical and biological assays.

  • High purity: 99.65%.
  • Molecular weight: 166.19 g/mol.
  • Formula: C8H15NaO2.
  • Suitable for HDAC inhibition and neuropharmacology research.
  • Available in laboratory-friendly quantities, including 25 g.

Catalog No. 50-002-44237 Supplier Medchemexpress LLC Supplier No. HY10585A25G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valproic acid sodium (sodium valproate) is the sodium salt of valproic acid for research use. It acts as an HDAC inhibitor and anticonvulsant, and is commonly used in studies of epilepsy, bipolar disorder, neuroprotection, and epigenetic regulation. Supplied as a solid powder suitable for dissolution for chemical and biological assays.

  • High purity: 99.65%.
  • Molecular weight: 166.19 g/mol.
  • Formula: C8H15NaO2.
  • Suitable for HDAC inhibition and neuropharmacology research.
  • Available in laboratory-friendly quantities, including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.