Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium valproate, 99.9%, MP Biomedicals™

Catalog No. ICN15206425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15206425 25 g
ICN15206410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15206425 Supplier MP Biomedicals, Inc Supplier No. 0215206425
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1069-66-5 | Formula: C8H15NaO2 | Weight: 166.196 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.196
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
Synonym sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
PubChem CID 16760703
ChEBI CHEBI:9925
IUPAC Name sodium;2-propylpentanoate
SMILES CCCC(CCC)C(=O)[O-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 0.999
Formula Weight 166.2
Percent Purity 100.6%
Grade Research
Chemical Name or Material Sodium valproate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.