Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Valproate, EP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18606904 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-904 25 g Amber Glass Bottle
18-606-905 100 g Amber Glass Bottle
18-606-906 500 g Amber Glass Bottle
18-606-907 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-904 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S247925GM
Add to Cart
Add to Cart

S2479, 1069-66-5, C8H15NaO2

Spectrum™ Chemical Sodium Valproate, EP is an anticonvulsant drug. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
IUPAC Name sodium 2-propylpentanoate
SMILES [Na+].CCCC(CCC)C([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.02
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98.5 to 101%
Grade EP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.