Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Valproate, EP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18606907 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-907 2.5 kg Poly Pail
18-606-904 25 g Amber Glass Bottle
18-606-905 100 g Amber Glass Bottle
18-606-906 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-907 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S24792.5KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S2479, 1069-66-5, C8H15NaO2

Spectrum™ Chemical Sodium Valproate, EP is an anticonvulsant drug. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
IUPAC Name sodium 2-propylpentanoate
SMILES [Na+].CCCC(CCC)C([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.02
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 98.5 to 101%
Grade EP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.