Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Valproate , MP Biomedicals, LLC

Catalog No. MP215206480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP215206480 100 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215206480 Supplier MP Biomedicals, Inc Supplier No. 0215206480
Add to Cart
Add to Cart

Sodium Valproate is a branched chain fatty acid which is reported to enhance central GABAergic neurotransmission and inhibit Na+ channels.

  • Sodium Valproate is reported to cause inositol depletion, activate the ERK pathway, inhibit GSK-3 α and GSK-3β
  • Valproic Acid has been reported to be a potent inhibitor of HDAC (histone deacetylase) in vitro (IC50 = 400μM for HDAC1), thereby relieving HDAC-dependent transcriptional repression and causes the hyperacetylation of histones in cultured cells
  • In animal studies, Valproic Acid has been observed to reduce tumor growth and metastasis formation
  • Valproic Acid is reported to activate Wnt-dependent gene expression and to mimic trichostatin A in the inhibition of histone deacetylase
  • This compound is also an inhibitor of the CYP2C9 enzyme
  • Anti-convulsant that also has efficacy as a mood stabilizer in bipolar disorder
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
Synonym sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
PubChem CID 16760703
ChEBI CHEBI:9925

Specifications

Color White
Packaging Glass
Quantity 100 g
Percent Purity 100.8
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.