Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pfaltz & Bauer SODIUM VINYL SULFONATE 25% AQ

Catalog No. 507490812
Encompass

Sodium Vinyl Sulfonate 25% Aq; 100g; CAS: 3039-83-6

Catalog No. 50-749-0812 Supplier Pfaltz & Bauer Supplier No. S07630100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Vinyl Sulfonate 25% Aq; 100g; CAS: 3039-83-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.