Learn More
Medchemexpress LLC Sodium zirconium cyclosilicate | 17141-74-1 | 95.0% | 100 MG
SDP
Catalog No. 5000233043
Encompass_Preferred
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate compound used as a selective cation-exchange reagent that binds potassium ions for biomedical and kidney disease research.
  • Selectively binds potassium ions for removal in vitro or preclinical studies.
  • Inorganic, non-absorbed microporous structure suitable for research applications.
  • Available in multiple small-scale sizes for laboratory use.
  • Supplied as a solid reagent with specified purity for experimental reproducibility.
Catalog No. 50-002-33043 Supplier Medchemexpress LLC Supplier No. HY119572100MG
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate compound used as a selective cation-exchange reagent that binds potassium ions for biomedical and kidney disease research.
  • Selectively binds potassium ions for removal in vitro or preclinical studies.
  • Inorganic, non-absorbed microporous structure suitable for research applications.
  • Available in multiple small-scale sizes for laboratory use.
  • Supplied as a solid reagent with specified purity for experimental reproducibility.